Orders LinkContactUsLinkComponent
Cables
Cables
InLab® cable MultiPin-BNC/RCA, 5 m
Katalog # 661-0251
Leverandør:   Mettler - Toledo
Cables
Katalog # 661-0251
Leverandør:  Mettler - Toledo
Cables
Cables
InLab® cable MultiPin-BNC/RCA, 5 m
Price
1.850,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Documents
Specifikationer
  • Beskrivelse:
    InLab® cable MultiPin-BNC/RCA
  • Længde:
    5 m
  • Seq:
    152