LiChrospher®, HPLC Columns
Analytisk, 100Diol, 100 Å, 5 µm
Katalog # LISPOH-5-10CE5
Leverandør: Avantor
Gå til:
Product Details & Documents
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
- Spherical porous silica
- Base deactivated LiChrospher® RP-select B
- High efficiency packed columns
Specifikationer
- Kolonne form:Analytisk
- Partikelstørrelse:5 µm
- Porestørrelse:100 Å
- Seq:61
- Fase:100Diol
- Cat. No.:LISPOH-5-10CE5
- Miljøcertificering:ISO 14001:2015 (530840 UM15)
- Environmentally Preferable:Lav fremstillingspåvirkning