Orders LinkContactUsLinkComponent
LiChrospher®, HPLC Columns
Si 60
Katalog # LISP60-5-250P
Leverandør:   Avantor
LiChrospher®, HPLC Columns
Katalog # LISP60-5-250P
Leverandør:  Avantor
LiChrospher®, HPLC Columns
LiChrospher®, HPLC Columns
Si 60
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
  • Spherical porous silica
  • Base deactivated LiChrospher® RP-select B
  • High efficiency packed columns
Specifikationer
  • Fase:
    Si 60