LiChrospher®, HPLC Columns
Si 60
Katalog # LISP60-5-250P
Leverandør: Avantor
Gå til:
Product Details & Documents
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
- Spherical porous silica
- Base deactivated LiChrospher® RP-select B
- High efficiency packed columns
Specifikationer
- Fase:Si 60