LiChrospher®, HPLC Columns
Analytisk, Si 60, 60 Å, 5 µm
Katalog # LISP60-5-10CP3
Leverandør: Avantor
Gå til:
Product Details & Documents
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
- Spherical porous silica
- Base deactivated LiChrospher® RP-select B
- High efficiency packed columns
Specifikationer
- Kolonne form:Analytisk
- Partikelstørrelse:5 µm
- Porestørrelse:60 Å
- Seq:2
- Fase:Si 60
- Cat. No.:LISP60-5-10CP3
- Miljøcertificering:ISO 14001:2015 (530840 UM15)
- Environmentally Preferable:Lav fremstillingspåvirkning