Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Accessories for centrifuges

Adapter (reducer), boring Ø×L: 30×77 mm, for 1× 50 ml tube (29×115 mm), for use with carrier 1560 in rotor 1554
  521-2224
   Lav fremstillingspåvirkning
 :   Hettich
Accessories for centrifuges
  521-2224
 :  Hettich
   Lav fremstillingspåvirkning
Accessories for centrifuges
Accessories for centrifuges

Adapter (reducer), boring Ø×L: 30×77 mm, for 1× 50 ml tube (29×115 mm), for use with carrier 1560 in rotor 1554

 
569,00 kr
   
1 * 1 Items
 
 
  • Beskrivelse:
    Adapter (reducer), boring Ø×L: 30×77 mm, for 1× 50 ml tube (29×115 mm), for use with carrier 1560 in rotor 1554
  • Seq:
    10
  • Miljøcertificering:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Environmentally Preferable:
    Lav fremstillingspåvirkning
  • Cat. No.:
    521-2224
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.