Storage Bags
Food storage bag, 180×340 mm
Katalog # HKPK100256
Leverandør: HK PACK KRAUTSCHEID
Gå til:
Product Details & Documents
LDPE transparent bags, ideal for storing and protecting food and small spare parts of all kinds.
- Food safety is ensured
- Flexible and tear resistant
- Freeze-resistant starting 50 μ and heat resistant up to 80 °C
Specifikationer
- Beskrivelse:Food storage bag
- Dimensioner:180×340 mm
- Seq:04