Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
UltraSpeed Pro Mopping Sysytem

UltraSpeed Pro Mopping Sysytem

UltraSpeed Pro Starter kit, single bucket
Catalog # VILE147184
Supplier:   FHP VILEDA SCS
UltraSpeed Pro Mopping Sysytem
Catalog # VILE147184
Supplier:  FHP VILEDA SCS
UltraSpeed Pro Mopping Sysytem
UltraSpeed Pro Mopping Sysytem

UltraSpeed Pro Starter kit, single bucket

Price
1.240,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Developed for all general floor cleaning tasks. All systems feature a simple to use press, integrated handle holders and an easy to manoeuvre chassis.

  • The fastest bucket and press microfibre mopping system
  • Able to clean over 750 m² per hour in a single step or 360 m² two step process
  • Fast wringing - simple to use and twice as fast
  • Efficient press - releases up to 50% more water
  • Floors dry faster and reduce slip hazards
  • Hygienic and no hand contact with mops or cleaning solution
  • Easy to use - no training required

To simplify the choice we have created three kits:
1. System : contains the mopping system; chassis, bucket and press.
2. Starter : containing the system plus mop frame and mop pad.
3. Ready to Go: containing the starter kit plus a telescopic mop handle.

Additionally if want to fulfil more cleaning tasks you can add the optional second bucket to the chassis to carry chemicals, cloths, paper etc. An ergonomic push bar is also available and all kits can be used with a choice of microfibre mops as well as specialist mops for cleaning safety floors.

Ergonomic and lightweight, no bending to wring, starter kits contain mop system, wringer, mop frame and mop pad. Ready to Go kits contain all items of the starter kit plus a telescopic handle. Ready to Go Kit Push contain all items of the Ready to Go kits plus a push bar.

Specifications
  • Beskrivelse:
    UltraSpeed Pro Starter kit, single bucket
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Beskrivelse
    Availability
    Price
  • VILE147597
    UltraSpeed Pro Wheel
    Log in for availability
    1 * 1 Items - 50,90 kr

Product Information

  • Catalog #
    Beskrivelse
    Availability
    Price
  • VILE147596
    UltraSpeed Pro Chassis only
    Log in for availability
    1 * 1 Items - 196,00 kr