Accessories for centrifuges
Carrier for swing-out rotor 1554, max. 4×200 ml, with single-hand clamp lock (without lid)
Catalog # 521-2220
Supplier: Hettich
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Carrier for swing-out rotor 1554, max. 4×200 ml, with single-hand clamp lock (without lid)
- Seq:49
- Cat. No.:521-2220
- Miljøcertificering:ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
- Environmentally Preferable:Lav fremstillingspåvirkning