Orders LinkContactUsLinkComponent
Tank liners, Corning®
Tank liners, Corning®
560 L, Material2-21408797|Material2-21408396, — mm
Katalog # MDTC91-300-60
Leverandør:   Corning
Tank liners, Corning®
Katalog # MDTC91-300-60
Leverandør:  Corning
Tank liners, Corning®
560 L, Material2-21408797|Material2-21408396, — mm
Price
3.940,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Corning® tank liners are designed to fit cylindrical tanks.
  • Available in a range of sizes
Tank liners are manufactured from LDPE.
Specifikationer
  • Diameter×H:
    — mm
  • Kapacitet:
    560 L
  • Seq:
    06
  • Slangemateriale:
    Material2-21408797|Material2-21408396