Orders LinkContactUsLinkComponent
VWR®, Dropping Bottles
VWR®, Dropping Bottles
Pipette kit for 20 ml dropping bottles, -
Katalog # 215-3825
Leverandør:   VWR Collection
VWR®, Dropping Bottles
Katalog # 215-3825
Leverandør:  VWR Collection
VWR®, Dropping Bottles
VWR®, Dropping Bottles
Pipette kit for 20 ml dropping bottles, -
Price
531,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Soda-lime glass, amber.
  • Good chemical and physical properties
  • Pipette kit not included
Amber to protect light-sensitive substances, with DIN 18 thread.
Specifikationer
  • Kapacitet:
    -
  • Beskrivelse:
    Pipette kit for 20 ml dropping bottles
  • Seq:
    10