Biotin hydrazide ≥95%
250 mg
Katalog # CAYM18418-250
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Biotin hydrazide is a biotinyl derivative used to label surface functional groups, antibodies, lectins, sugars, nucleic acids, or molecules with free carboxylic or keto groups.
Used as a probe for determining protein carbonylation, an irreversible posttranslational modification resulting from the actions of reactive oxygen species or lipid oxidation products. The reaction of this probe is direct and does not require catalysts or reducing agents.
Synonymer: 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide
- CAS nummer:66640-86-6
- Formel:C₁₀H₁₈N₄O₂S
- MDL nummer:MFCD00078532
- Molecular Weight:258.34 g/mol
Specifikationer
Specifikationstestresultater
- Purity:≥95%
- Formulation:A Crystalline Solid
Specifikationer
- Pakn:250 mg



