Orders LinkContactUsLinkComponent
GC guard columns, HydroGOLD
GC guard columns, HydroGOLD
Forkolonne, 0,25 mm, 5 m
Katalog # 555-9121
Leverandør:   Thermo Fisher Scientific
GC guard columns, HydroGOLD
Katalog # 555-9121
GC guard columns, HydroGOLD
Forkolonne, 0,25 mm, 5 m
Price
895,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These guard columns extend analytical column lifetime by preventing degradation from harsh 'steam cleaning' water injections.
  • Highly deactivated to provide superior inertness, essential for analysis of active compounds
  • Protects against column contamination caused by non-volatile materials
Specifikationer
  • Kolonne form:
    Forkolonne
  • Ø indv.:
    0,25 mm
  • Længde:
    5 m
  • Seq:
    06