GC guard columns, HydroGOLD
Forkolonne, 0,53 mm, 2 m
Katalog # 555-9123
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
These guard columns extend analytical column lifetime by preventing degradation from harsh 'steam cleaning' water injections.
- Highly deactivated to provide superior inertness, essential for analysis of active compounds
- Protects against column contamination caused by non-volatile materials
Specifikationer
- Kolonne form:Forkolonne
- Ø indv.:0,53 mm
- Længde:2 m
- Seq:04