Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Lens cleaning tissues

Lens cleaning tissue, 80×100 mm
Katalog # BINZ146099
Leverandør:   Ahlstrom
Lens cleaning tissues
Katalog # BINZ146099
Leverandør:  Ahlstrom
Lens cleaning tissues
Lens cleaning tissues

Lens cleaning tissue, 80×100 mm

Price
18,10 kr
Log ind for tilgængelighed
1 * 25 Items
Product Details & Documents

These very fine lens cleaning tissues are made of pure manila vegetable fibre.

  • Non abrasive

Lint-free paper for cleaning of optical surfaces such as lenses and mirrors of microscopes, etc.

12 g/m²

Specifikationer
  • Beskrivelse:
    Lens cleaning tissue
  • L×B:
    80×100 mm
  • Seq:
    01