Orders LinkContactUsLinkComponent
Pipette leak testing unit
PLTconnect software for PLT-unit
Katalog # 613-5740
Leverandør:   Brand
Pipette leak testing unit
Katalog # 613-5740
Leverandør:  Brand
Pipette leak testing unit
Pipette leak testing unit
PLTconnect software for PLT-unit
Price
1.410,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The PLT (Pipette Leak Tester) unit for air displacement pipettes allows daily pipette checks, providing a safeguard during periods between routine calibrations thereby significantly improving the pipette process reliability. Even the smallest leaks are detected and quantified.
  • Test limit values for all commercially available single- and multi-channel pipettes in the volume range of 1 μl to 10 ml are pre-programmed
  • Test pipettes with and without tips to assist in diagnosis of the leak source
  • Test results in seconds
  • Optional PLT unit software (PLTconnect) facilitates the documentation of leak tests
 : Supplied with 1 of each type of adapter (4-channel pipette adapter optional) for testing air displacement pipettes with tip (attached) or testing without tip, 2 plugs (for instrument validation), 3 replacement PE filters for the pipette adapters, universal AC adapter, quality certificate and operating manual.
Specifikationer
  • Beskrivelse:
    PLTconnect software for PLT-unit
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Filter for PLT unit pipette adapter
    Filter for PLT unit pipette adapter
    613-1693
    Log ind for tilgængelighed
    1 * 10 Items - 66,80 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Pipette adapter for PLT unit, 1-channel, with tip
    Pipette adapter for PLT unit, 1-channel, with tip
    613-1690
    Log ind for tilgængelighed
    1 * 1 Items - 383,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Pipette adapter for PLT unit, 1-channel, without tip
    Pipette adapter for PLT unit, 1-channel, without tip
    613-1691
    Log ind for tilgængelighed
    1 * 1 Items - 383,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Pipette adapter for PLT unit, 4-channel
    Pipette adapter for PLT unit, 4-channel
    613-1692
    Log ind for tilgængelighed
    1 * 1 Items - 1.240,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Universal AC adapter, 100 - 240 V; 50/60 Hz
    Universal AC adapter, 100 - 240 V; 50/60 Hz
    613-1694
    Log ind for tilgængelighed
    1 * 1 Items - 600,00 kr