Calciumstearat NF
Fiber drum, 45 kg
Katalog # SPCMCA169-45KGBL
Leverandør: Spectrum Chemical
Gå til:
Product Details & Documents
BSE/TSE statement (available upon request)
Kosher certified
Synonymer: Stearic acid calcium salt
- CAS nummer:1592-23-0
- Formel:C₃₆H₇₀CaO₄
- MDL nummer:MFCD00036390
- Molecular Weight:607.03 g/mol
- Merck Index:13,01708
- Smeltende Pt:147…149 °C
- Tæthed:1,065 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:216-472-8
Specifikationer
Specifikationstestresultater
- Assay:9% Min.
Specifikationer
- Pakning:Fiber drum
- Pakn:45 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



