Calciumstearat NF
Poly pail, 2,5 kg
Katalog # SPCMCA169-2.5KG
Leverandør: Spectrum Chemical
Gå til:
Product Details & Documents
BSE/TSE statement (available upon request)
Kosher certified
Synonymer: Stearic acid calcium salt
- CAS nummer:1592-23-0
- Formel:C₃₆H₇₀CaO₄
- MDL nummer:MFCD00036390
- Molecular Weight:607.03 g/mol
- Merck Index:13,01708
- Smeltende Pt:147…149 °C
- Tæthed:1,065 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:216-472-8
Specifikationer
Specifikationstestresultater
- Assay:9% Min.
Specifikationer
- Pakning:Poly pail
- Pakn:2,5 kg



