Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-ɑ-Tocopherol succinate ≥96.0% USP

D-ɑ-Tocopherol succinate ≥96.0% USP

25 kg
Katalog # SPCMV1035-25KGBL
Leverandør:   Spectrum Chemical
D-ɑ-Tocopherol succinate ≥96.0% USP
Katalog # SPCMV1035-25KGBL
Leverandør:  Spectrum Chemical
CAS nummer:  4345-03-3
D-ɑ-Tocopherol succinate ≥96.0% USP

25 kg

CAS nummer:  4345-03-3
Price
60.230,00 kr
Log ind for tilgængelighed
1 * 25 kg
Product Details & Documents

RRR-Vitamin E Succinate, 1 mg = 0.81 alpha-tocopherol equivalents, USP.

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonymer:  Vitamin E Succinate, D-α-Tocopherol Succinate, 4-Oxo-4-(((R)-2,5,7,8-tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoic acid
  • CAS nummer:4345-03-3
  • Formel:C₃₃H₅₄O₅
  • MDL nummer:MFCD00072055
  • Molecular Weight:530.79 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationstestresultater

  • Assay:
    96.0 - 102.0%
  • Acidity:
    To Pass Test
  • Identification:
    To Pass Test

Specifikationer

  • Pakn:
    25 kg

Specifikationer