D-ɑ-Tocopherol succinate ≥96.0% USP
Glass, amber, 1 kg
Katalog # SPCMV1035-1KG
Leverandør: Spectrum Chemical
D-ɑ-Tocopherol succinate ≥96.0% USP
Katalog # SPCMV1035-1KG
Leverandør: Spectrum Chemical
CAS nummer: 4345-03-3
Jump to:
Product Details & Documents
RRR-Vitamin E Succinate, 1 mg = 0.81 alpha-tocopherol equivalents, USP.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Synonyms: Vitamin E Succinate, D-α-Tocopherol Succinate, 4-Oxo-4-(((R)-2,5,7,8-tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoic acid
- CAS Number:4345-03-3
- Formula:C₃₃H₅₄O₅
- MDL Number:MFCD00072055
- Molecular Weight:530.79 g/mol
- Storage Temperature:Stue temperatur
Specifications
Specification Test Results
- Assay:96.0 - 102.0%
- Acidity:To Pass Test
- Identification:To Pass Test
Specifications
- Pakning:Glass, amber
- Pakn:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



