Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tannin USP

Tannin USP

Poly pail, 2,5 kg
Katalog # SPCMT1013-2.5KG
Leverandør:   Spectrum Chemical
Tannin USP
Katalog # SPCMT1013-2.5KG
Leverandør:  Spectrum Chemical
CAS nummer:  1401-55-4
Tannin USP

Poly pail, 2,5 kg

CAS nummer:  1401-55-4
Price
7.240,00 kr
Log ind for tilgængelighed
1 * 2,5 kg
Product Details & Documents
Synonymer:  Gallotannin, Tannin
  • CAS nummer:1401-55-4
  • Formel:C₇₆H₅₂O₄₆
  • MDL nummer:MFCD00066397
  • Molecular Weight:1701.2 g/mol
  • Merck Index:12,09221
  • Smeltende Pt:218…219 °C
  • Tæthed:>1 g/cm³ (20 °C)
  • Flash Pt:199 °C
  • EINECS:215-753-2
Specifikationer

Specifikationer

  • Pakning:
    Poly pail
  • Pakn:
    2,5 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.