Sorbitanmonostearat NF
Poly pail, 12 kg
Katalog # SPCMSPA63-12KG
Leverandør: Spectrum Chemical
Gå til:
Product Details & Documents
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonymer: Sorbitan monooctadecanoate, (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol monooctadecanoate, Sorbitan stearate
- CAS nummer:1338-41-6
- Formel:C₂₄H₄₆O₆
- MDL nummer:MFCD00005366
- Molecular Weight:430.63 g/mol
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-664-9
Specifikationer
Specifikationstestresultater
- Assay:68% Min.
Specifikationer
- Pakning:Poly pail
- Pakn:12 kg



