Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Brij® S100

Brij® S100

Glass, amber, 2,5 kg
Katalog # SPCMB1593-2.5KG
Leverandør:   Spectrum Chemical
Brij® S100
Katalog # SPCMB1593-2.5KG
Leverandør:  Spectrum Chemical
CAS nummer:  9005-00-9
Brij® S100

Glass, amber, 2,5 kg

CAS nummer:  9005-00-9
Price
5.720,00 kr
Log ind for tilgængelighed
1 * 2,5 kg
Product Details & Documents
Synonymer:  Polyoxyethylene (100) stearyl ether
  • CAS nummer:9005-00-9
  • Formel:C₁₈H₃₇(OCH₂CH₂)nOH,n~100
  • MDL nummer:MFCD00074177
  • EINECS:500-017-8
Specifikationer

Specifikationer

  • Pakning:
    Glass, amber
  • Pakn:
    2,5 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.