Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Allantoin ≥98.5% USP

Allantoin ≥98.5% USP

25 kg
Katalog # SPCMA2721-25KGBL
Leverandør:   Spectrum Chemical
Allantoin ≥98.5% USP
Katalog # SPCMA2721-25KGBL
Leverandør:  Spectrum Chemical
CAS nummer:  97-59-6
Allantoin ≥98.5% USP

25 kg

CAS nummer:  97-59-6
Price
33.460,00 kr
Log ind for tilgængelighed
1 * 25 kg
Product Details & Documents

Allantoin, USP is a moisturising ingredient.

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonymer:  1-(2,5-Dioxoimidazolidin-4-yl)urea
  • CAS nummer:97-59-6
  • Formel:C₄H₆N₄O₃
  • MDL nummer:MFCD00005260
  • Molecular Weight:158.12 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationstestresultater

  • Assay:
    98.5 - 101.0%
  • Acidity or Alkalinity:
    To Pass Test
  • Loss On Drying:
    max. 0.1 %
  • Organic Impurities:
    max. 0.5 %
  • Residue On Ignition (Sulphated Ash):
    max. 0.1 %

Specifikationer

  • Pakn:
    25 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.