L(+)-Leucin USP
Poly pail, 5 kg
Katalog # SPCML1202-5KG
Leverandør: Spectrum Chemical
Gå til:
Product Details & Documents
Synonymer: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
- CAS nummer:61-90-5
- Formel:C₆H₁₃NO₂
- MDL nummer:MFCD00002617
- Molecular Weight:131.17 g/mol
- Merck Index:14,05451
- Smeltende Pt:300 °C
- Tæthed:1,165 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-522-0
Specifikationer
Specifikationstestresultater
- Assay:98.5% Min.
Specifikationer
- Pakning:Poly pail
- Pakn:5 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



