Hydroquinon USP
Poly pail, 2,5 kg
Katalog # SPCMHY113-2.5KG
Leverandør: Spectrum Chemical
Gå til:
Product Details & Documents
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonymer: 1,4-Dihydroxybenzen, Benzen-1,4-diol
- CAS nummer:123-31-9
- Formel:C₆H₄(OH)₂
- MDL nummer:MFCD00002339
- Molecular Weight:110.11 g/mol
- Merck Index:13,04833
- Kogende Pt:285 °C (1013 hPa)
- Smeltende Pt:172 °C
- Tæthed:1,332 g/cm³ (20 °C)
- Flash Pt:165 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-617-8
- FN:3077
- ADR:9,III
Specifikationer
Specifikationstestresultater
- Assay:99% Min.
Specifikationer
- Pakning:Poly pail
- Pakn:2,5 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



