L-Alanin USP
Poly pail, 5 kg
SPCMA1253-5KG
: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
- :56-41-7
- :C₃H₇NO₂
- :MFCD00064410
- :89.09 g/mol
- :13,00203
- :189 °C (1013 hPa)
- :79 °C
- :1,4 g/cm³ (20 °C)
- :Stue temperatur
- :200-273-8
- Assay:98.5% Min.
- Pakning:Poly pail
- Pakn:5 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



