L-Alanin USP
Glass, amber, 1 kg
SPCMA1253-1KG
: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
- :56-41-7
- :C₃H₇NO₂
- :MFCD00064410
- :89.09 g/mol
- :13,00203
- :189 °C (1013 hPa)
- :79 °C
- :1,4 g/cm³ (20 °C)
- :Stue temperatur
- :200-273-8
- Assay:98.5% Min.
- Pakning:Glass, amber
- Pakn:1 kg



