Cyclopentanone ≥99%
200 L
Katalog # SPCMC1124-200LTBL
Leverandør: Spectrum Chemical
Gå til:
Product Details & Documents
Cyclopentanone is a liquid that has an odor similar to peppermint and is devoid of color. It's an organic compound and a cyclic ketone that most often used as a fragrance.
GHS hazard : Flame;Exclamation mark
UN2245
- CAS nummer:120-92-3
- Formel:C₅H₈O
- MDL nummer:MFCD00001409
- Molecular Weight:84.12 g/mol
- Tæthed:0,951
- Opbevaringstemperatur:Stue temperatur
- FN:2245
- ADR:3,III
Specifikationer
Specifikationstestresultater
- Assay:99% min.
Specifikationer
- Pakn:200 L
- Cat. No.:SPCMC1124-200LTBL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



