Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Testkits, sulfat test, til Spectroquant®, Supelco®

Sulphate test, photometric, 0,5 - 50 mg/L SO42−, Barium sulphate, turbidimetric, Move 100, Nova 60 A, Prove 100 (plus), Prove 300 (plus), Prove 600 (plus), 1 L
Katalog # 1.01812.0001
Leverandør:   Merck
Testkits, sulfat test, til Spectroquant®, Supelco®
Katalog # 1.01812.0001
Leverandør:  Merck
CAS nummer:  
Testkits, sulfat test, til Spectroquant®, Supelco®
Testkits, sulfat test, til Spectroquant®, Supelco®

Sulphate test, photometric, 0,5 - 50 mg/L SO42−, Barium sulphate, turbidimetric, Move 100, Nova 60 A, Prove 100 (plus), Prove 300 (plus), Prove 600 (plus), 1 L

CAS nummer:  
Price
1.100,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents

All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.

  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times

Spectroquant® reagent test kits

Highly stable, ready to use reagent mixtures in a user friendly design.
Free selection of appropriate cell format according to the measuring range.
Long shelf life up to 3 years all at room temperature.
Automatic selection of the correct method with help of AutoSelector barcode identification.

Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.

Specifikationer

Specifikationer

  • Kemisk metode:
    Barium sulphate, turbidimetric
  • Pakn:
    1 L
  • Beskrivelse:
    Sulphate test, photometric
  • Til enhed:
    Move 100, Nova 60 A, Prove 100 (plus), Prove 300 (plus), Prove 600 (plus)
  • Område:
    0,5 - 50 mg/L SO<sub>4</sub><sup>2−</sup>
  • Cat. No.:
    1.01812.0001
  • M-Clarity quality level:
    MQ100

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.