pH and ORP electrodes, IDS, wireless ready
IDS pH electrode, SenTix® Micro 900-P, wireless ready, SenTix® Micro 900-P, 0 - 14, 0...+100 °C, Glas, KCl 3 mol/l, Platinum wire, 12/5×70/130 mm, Plug head for AS IDS cable or wireless module
Katalog # 662-2129
Leverandør: WTW
Gå til:
Product Details & Documents
High quality, enhanced sensor technology combined with most modern measurement electronics. The electrodes feature a plug head which can either be connected to an IDS cable or to a wireless module.
- Intelligent: Electrodes are automatically recognized by the meter, each sensor is exactly identified
- Digital: Measurement signals are processed digitally, providing very accurate data
- Measurement integrity benchmarking of the electrodes by QSC (Quality Sensor Control)
- IDS electrodes store their serial number and their calibration records
- Wireless ready: Disturbing cables are no longer required
Ordering information: Electrodes are supplied with plug head but without cable or wireless module which need to be ordered separately.
Documents
Specifikationer
- Tilslutning:Plug head for AS IDS cable or wireless module
- Ø×L:12/5×70/130 mm
- Model:SenTix® Micro 900-P
- Beskrivelse:IDS pH electrode, SenTix® Micro 900-P, wireless ready
- Materiale:Glas
- Seq:14
- Membran:Platinum wire
- Elektrolyt:KCl 3 mol/l
- Temperatur:0...+100 °C
- pH:0 - 14
Accessories
Produktinformation
- Katalog #LængdeTilgængelighedPrice
- AS/IDS-1,5, connection cable for IDS sensors with waterproof plug headAS/IDS-1,5, connection cable for IDS sensors with waterproof plug head662-17331,5 mLog ind for tilgængelighed1 * 1 Items - 1.160,00 kr
Produktinformation
- Katalog #LængdeTilgængelighedPrice
- AS/IDS-3, connection cable for IDS sensors with waterproof plug headAS/IDS-3, connection cable for IDS sensors with waterproof plug head662-17343 mLog ind for tilgængelighed1 * 1 Items - 1.580,00 kr
Produktinformation
- Katalog #LængdeTilgængelighedPrice
- AS/IDS-10, connection cable for IDS sensors with waterproof plug headAS/IDS-10, connection cable for IDS sensors with waterproof plug head662-173610 mLog ind for tilgængelighed1 * 1 Items - 1.990,00 kr
Produktinformation
- Katalog #LængdeTilgængelighedPrice
- AS/IDS-15, connection cable for IDS sensors with waterproof plug headAS/IDS-15, connection cable for IDS sensors with waterproof plug head662-173715 mLog ind for tilgængelighed1 * 1 Items - 2.280,00 kr
Produktinformation
- Katalog #LængdeTilgængelighedPrice
- AS/IDS-20, connection cable for IDS sensors with waterproof plug headAS/IDS-20, connection cable for IDS sensors with waterproof plug head662-173820 mLog ind for tilgængelighed1 * 1 Items - 2.490,00 kr
Produktinformation
- Katalog #LængdeTilgængelighedPrice
- AS/IDS-40, connection cable for IDS sensors with waterproof plug headAS/IDS-40, connection cable for IDS sensors with waterproof plug head662-174040 mLog ind for tilgængelighed1 * 1 Items - 7.140,00 kr
Produktinformation
- Katalog #LængdeTilgængelighedPrice
- AS/IDS-60, connection cable for IDS sensors with waterproof plug headAS/IDS-60, connection cable for IDS sensors with waterproof plug head662-174160 mLog ind for tilgængelighed1 * 1 Items - 8.590,00 kr
Produktinformation
- Katalog #LængdeTilgængelighedPrice
- AS/IDS-100, connection cable for IDS sensors with waterproof plug headAS/IDS-100, connection cable for IDS sensors with waterproof plug head662-1742100 mLog ind for tilgængelighed1 * 1 Items - 11.160,00 kr