Beakers, with handle, PP, Corning®
1000 ml, 167 mm, 125 mm, 10 ml
Leverandør: Corning
Gå til:
Product Details & Documents
PP, translucent.
- Smooth surfaces for easy and fast cleaning
- Printed with blue graduations
- With handle and spout
Ideal for general laboratory use and a wide variety of applications.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Udv.Ø:125 mm
- Inddelings-interval:10 ml
- Kapacitet:1000 ml
- Højde:167 mm