Multiscreen® High Volume Filter Plates
MultiScreen® High volume filter plates, PP, Opaque, Hydrophilic Durapore®
membrane, 0,45 µm, 96, +
membrane, 0,45 µm, 96, +
Katalog # MILFMVHVN4525
Leverandør: Merck
Gå til:
Product Details & Documents
The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.
- Sample prep
- Protein kinase/phosphatase assays
- Receptor/ligand binding assays
Specifikationer
- Membran:Hydrophilic Durapore®
membrane - Ant. brønde:96
- Porestørrelse:0,45 µm
- Steril:+
- Farve:Opaque
- Beskrivelse:MultiScreen® High volume filter plates
- Materiale:PP
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



