Orders LinkContactUsLinkComponent
Bag cutters
Bag cutter
Katalog # MICRBC-01
Leverandør:   MICRONOVA
Bag cutters
Katalog # MICRBC-01
Leverandør:  MICRONOVA
Bag cutters
Bag cutters
Bag cutter
Price
647,00 kr
Log ind for tilgængelighed
1 * 6 Items
Product Details & Documents
Plastic cutters for opening raw material packages.
  • Automatically retractable safety blade guard
  • No blade changes required - eliminates all loose blades
  • Ergonomic handle
  • Available gamma irradiated
Specifikationer
  • Beskrivelse:
    Bag cutter
  • Seq:
    01