Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Threaded fittings

Threaded fittings

Socket, inner thread, PP, 12,7 mm, nominal width 19 mm, PP, 12,7 mm
Katalog # BURK8501-0012
Leverandør:   BURKLE
Threaded fittings
Katalog # BURK8501-0012
Leverandør:  BURKLE
Threaded fittings
Threaded fittings

Socket, inner thread, PP, 12,7 mm, nominal width 19 mm, PP, 12,7 mm

Price
38,70 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Gevind:
    12,7 mm
  • Beskrivelse:
    Socket, inner thread, PP, 12,7 mm, nominal width 19 mm
  • Materiale:
    PP
  • Seq:
    42
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.