Orders LinkContactUsLinkComponent
pH buffer solutions
pH buffer solutions
Technical buffer solution, 30×20 ml, 11,00, 30
Katalog # METT30111135
Leverandør:   Mettler - Toledo
pH buffer solutions
Katalog # METT30111135
Leverandør:  Mettler - Toledo
CAS nummer:  
pH buffer solutions
pH buffer solutions
Technical buffer solution, 30×20 ml, 11,00, 30
CAS nummer:  
Price
715,00 kr
Log ind for tilgængelighed
1 * 30 Items
Product Details & Documents
Specifikationer
Specifikationer
  • Pakn:
    30
  • Beskrivelse:
    Technical buffer solution, 30×20 ml
  • pH-værdi:
    11,00
Specifikationer