Orders LinkContactUsLinkComponent
VWR® Coagulase Test
VWR® Coagulase Test
Coagulase test, 3 ml, 10 Ampoul
Katalog # 673333ZL
Leverandør:   VWR Chemicals
VWR® Coagulase Test
Katalog # 673333ZL
Leverandør:  VWR Chemicals
CAS nummer:  
VWR® Coagulase Test
VWR® Coagulase Test
Coagulase test, 3 ml, 10 Ampoul
CAS nummer:  
Price
1.830,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Confirmative test for coagulase-positive Staphylococcus.
  • Formulated according to ISO 6888-1
  • Can be used to differentiate Staphylococcus aureus from coagulase-negative Staphylococci
Specifikationer
Specifikationer
  • Pakn:
    10 Ampoul
  • Beskrivelse:
    Coagulase test, 3 ml
  • Cat. No.:
    673333ZL
  • Shelf life:
    1080 days (Ampoule)
  • Opbevaringstemperatur:
    2...8 °C
Specifikationer