Chloroform ≥99.5% stabilised
Glass bottle, 2,5 L
Leverandør: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Katalog # C2432-2.5L
Leverandør: Honeywell Chemicals
CAS nummer: 67-66-3
Gå til:
Product Details & Documents
Beilstein Number: 1731042
RTECS Number: FS9100000
UNSPSC Code: 12191502
Synonymer: Trichlormethan
- CAS nummer:67-66-3
- Formel:CHCl₃
- MDL nummer:MFCD00000826
- Molecular Weight:119.38 g/mol
- Kogende Pt:61 °C (1,013 hPa)
- Smeltende Pt:–61,3 °C
- Smeltende Pt:-61,3 °C
- Tæthed:1,485 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-663-8
- FN:1888
- ADR:6.1,III
- :stabiliseret med amylen 100-200 ppm
Documents
Specifikationer
Specifikationstestresultater
- Vapor Pressure:160 mmHg ( 20 °C)
- NMR-spectrum:complying
- Assay (GC):Min. 99.50 %
- 2-methyl-2-buten (GC):20 - 200 ppm
- Refractive Index:n20/D 1.445 (lit.)
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



