Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Pipette controller, Pipet-Aid® XP2

Pipette controller, Pipet-Aid® XP2

Pipet-Aid® XP2, CE, with UK plug, 230 V, 1 - 100 ml
Katalog # DRUM4-000-501-UK
Leverandør:   DRUMMOND
Pipette controller, Pipet-Aid® XP2
Katalog # DRUM4-000-501-UK
Leverandør:  DRUMMOND
Pipette controller, Pipet-Aid® XP2

Pipet-Aid® XP2, CE, with UK plug, 230 V, 1 - 100 ml

Price
10.090,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

This controller is an advanced version of the Pipet-Aid® XP controller and accepts 1 to 100 ml pipettes.

  • New ergonomic design
  • New power source for uninterrupted extended operation; can be charged while in use
  • Ultra quiet precision pump provides great control for aspiration or dispensing

Leveringsinformation: The unit is supplied complete with a power supply/charger, four extra filters, and a removable stand.

Specifikationer
  • Kapacitet:
    1 - 100 ml
  • Beskrivelse:
    Pipet-Aid® XP2, CE, with UK plug, 230 V
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.