Orders LinkContactUsLinkComponent
Test strips, peroxide, Dosatest
Test strips, peroxide, Dosatest
Peroxide test strips 0 - 100 mg/l, 0 · 1 · 3 · 10 · 30 · 100, 100
Katalog # 85434.601
Leverandør:   VWR Chemicals
Test strips, peroxide, Dosatest
Katalog # 85434.601
Leverandør:  VWR Chemicals
CAS nummer:  
Test strips, peroxide, Dosatest
Test strips, peroxide, Dosatest
Peroxide test strips 0 - 100 mg/l, 0 · 1 · 3 · 10 · 30 · 100, 100
CAS nummer:  
Price
230,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
A ready to use kit that fits into your lab coat pocket. Intended for water-based samples
  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision
Ideal to monitor your desinfection processes. Semi-quantitative results can be obtained between 10 and 120 seconds.
The colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Emballering: Container with 100 test strips 6×95 mm.
Specifikationer
Specifikationer
  • Pakn:
    100
  • Beskrivelse:
    Peroxide test strips 0 - 100 mg/l
  • Område:
    0 · 1 · 3 · 10 · 30 · 100
Specifikationer