n-Hexan, anhydrous, over molecular sieves
1 L
Leverandør: Thermo Fisher Scientific
n-Hexan, anhydrous, over molecular sieves
Katalog # 47104.K2
Leverandør: Thermo Fisher Scientific
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
CAS No.: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Appearance: Colourless Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 UN No.: UN1208 MDL No.: MFCD00009520
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:14,4694
- Kogende Pt:69 °C
- Smeltende Pt:–95 °C
- Tæthed:0,659
- Flash Pt:–23 °C (–9 °F)
- Opbevaringstemperatur:Stue temperatur
- FN:1208
- ADR:3,II
- :Packaged under argon in resealable ChemSeal™ bottles.
- EINECS:203-777-6
Documents
Specifikationer
Specifikationer
- Pakn:1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



