n-Hexan, anhydrous, over molecular sieves
250 mL
Leverandør: Thermo Fisher Scientific
n-Hexan, anhydrous, over molecular sieves
Katalog # 47104.AK
Leverandør: Thermo Fisher Scientific
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
CAS No.: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Appearance: Colourless Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 UN No.: UN1208 MDL No.: MFCD00009520
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:14,4694
- Kogende Pt:69 °C
- Smeltende Pt:–95 °C
- Tæthed:0,659
- Flash Pt:–23 °C (–9 °F)
- Opbevaringstemperatur:Stue temperatur
- FN:1208
- ADR:3,II
- :Packaged under argon in resealable ChemSeal™ bottles.
- EINECS:203-777-6
Documents
Specifikationer
Specifikationer
- Pakn:250 mL