Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Super Adjustable™ Super Erecta® shelving

Super Adjustable™ Super Erecta® shelving

Super Adjustable™ wire shelf, A2142NS, Rustfrit stål, 1066×530×1370 mm, 5,4 kg
Katalog # METOA2142NS
Leverandør:   Metro International
Super Adjustable™ Super Erecta® shelving
Katalog # METOA2142NS
Leverandør:  Metro International
Super Adjustable™ Super Erecta® shelving
Super Adjustable™ Super Erecta® shelving

Super Adjustable™ wire shelf, A2142NS, Rustfrit stål, 1066×530×1370 mm, 5,4 kg

Price
4.970,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Designed to work in conjunction with the entire Super Erecta® System of shelves and accessories, these Super Adjustable wire shelves have a unique corner release system which allows them to be adjusted quickly and easily without the use of tools.

  • Heavy duty construction
  • Built to save space and to stand though against years of heavy use
  • Adjust and add shelves quickly and easily to reclaimwasted space

They shelf have a sleek chrome finish that makes it perfect for dry storage areas, while the open-wire construction minimises dust accumulation and allows a free circulation of air and greater visibility.

Specifikationer
  • Beskrivelse:
    Super Adjustable™ wire shelf, A2142NS
  • Materiale:
    Rustfrit stål
  • Seq:
    39
  • B×D×H:
    1066×530×1370 mm
  • Vægt:
    5,4 kg