Mobile storage cabinets
Mobile storage cabinet, lockable, 2 door, 900×516×740 mm
Katalog # KOTT315-00137
Leverandør: KOETTERMANN
Gå til:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifikationer
- Beskrivelse:Mobile storage cabinet, lockable
- Seq:31
- B×D×H:900×516×740 mm
- Døre:2 door