Mobile storage cabinets
Mobile storage cabinet, with 2 drawer of 150 mm height and 1 drawer of 300 mm height, lockable, 3 drawers, 900×516×740 mm
Katalog # KOTT315-00134
Leverandør: KOETTERMANN
Gå til:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifikationer
- Beskrivelse:Mobile storage cabinet, with 2 drawer of 150 mm height and 1 drawer of 300 mm height, lockable
- Seq:28
- B×D×H:900×516×740 mm
- Døre:3 drawers