Mobile storage cabinets
Mobile storage cabinet with 1 shelf, With front open, 450×500×740 mm
Katalog # KOTT315-00116
Leverandør: KOETTERMANN
Gå til:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifikationer
- Beskrivelse:Mobile storage cabinet with 1 shelf
- Seq:10
- B×D×H:450×500×740 mm
- Døre:With front open