Mobile storage cabinets
Mobile storage cabinet, with 4 drawer of 150 mm height, lockable, 4 drawers, 1200×516×740 mm
Katalog # KOTT315-00106
Leverandør: KOETTERMANN
Gå til:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifikationer
- Beskrivelse:Mobile storage cabinet, with 4 drawer of 150 mm height, lockable
- Seq:51
- B×D×H:1200×516×740 mm
- Døre:4 drawers