Orders LinkContactUsLinkComponent
Mobile storage cabinets
Mobile storage cabinet, 2 doors, 1200×516×740 mm
Katalog # KOTT315-00100
Leverandør:   KOETTERMANN
Mobile storage cabinets
Katalog # KOTT315-00100
Leverandør:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, 2 doors, 1200×516×740 mm
Price
6.500,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Epoxy powder coated steel cabinets
  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifikationer
  • Beskrivelse:
    Mobile storage cabinet
  • Seq:
    45
  • B×D×H:
    1200×516×740 mm
  • Døre:
    2 doors