Orders LinkContactUsLinkComponent
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, with 3 drawers of 150 mm height, lockable, 3 drawers, 1200×516×590 mm
Katalog # KOTT315-00094
Leverandør:   KOETTERMANN
Mobile storage cabinets
Katalog # KOTT315-00094
Leverandør:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, with 3 drawers of 150 mm height, lockable, 3 drawers, 1200×516×590 mm
Price
9.500,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Epoxy powder coated steel cabinets
  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifikationer
  • Beskrivelse:
    Mobile storage cabinet, with 3 drawers of 150 mm height, lockable
  • Seq:
    39
  • B×D×H:
    1200×516×590 mm
  • Døre:
    3 drawers