Mobile storage cabinets
Mobile storage cabinet, 2 door, 900×516×590 mm
Katalog # KOTT315-00131
Leverandør: KOETTERMANN
Gå til:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifikationer
- Beskrivelse:Mobile storage cabinet
- Seq:23
- B×D×H:900×516×590 mm
- Døre:2 door