Volumetric flasks, class A, white markings
25 ml, 14/23, 0,06 ± ml
Catalog # WITG3.670.018
Supplier: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3.3, amber, with glass stopper.
- White markings
- Conformity certified (DE-M marking)
- Inclusive batch certificate
: DIN EN ISO 1042
Specifikationer
- NS:14/23
- Kapacitet:25 ml
- Seq:06
- Tolerance:0,06 ± ml